Learn More
SAI Infusion Technologies Pre-filled Syringes - 10cc Syringe filled with 10ml Saline
SDP
Catalog No. NC0044250
Encompass
Pre-filled Syringes - 10cc Syringe filled with 10ml Saline, white button cap. Sterile prefilled syringes save time and money in preclinical studies requiring repeated catheter access. These 10cc syringes are pre-filled with 10ml of 0.9% Sodium Chloride, 30/case. Product is for laboratory animal research only.
Catalog No. NC0044250 Supplier SAI Infusion Technologies Supplier No. SC1010
Pre-filled Syringes - 10cc Syringe filled with 10ml Saline, white button cap. Sterile prefilled syringes save time and money in preclinical studies requiring repeated catheter access. These 10cc syringes are pre-filled with 10ml of 0.9% Sodium Chloride, 30/case. Product is for laboratory animal research only.