Learn More
American Biotech Supply 10CF PHARMACY REFRIG/FREEZER
Catalog No. NC2468995
Encompass_Preferred
5.2 CU. FT. CAPACITY (REFRIGERATOR) 4 CU. FT. CAPACITY (FREEZER) ONE GLASS SWING DOOR (FRIDGE)ONE SOLID SWING DOOR (FREEZER)3 ADJUSTABLE SHELVES (FREEZER) 3 ADJUSTABLE SHELVES (FRIDGE)BOTTOM FOOTED SHELF TO ALLOW AIR CIRCULATIONBACK WALL EVAPORATOR COVERKEYED DOOR LOCKSMAGNETIC DOOR GASKET FOR POSITIVE SEALRECESSED DOOR HANDLESSTACKING KITLEVELING LEGSLED INTERIOR LIGHTPROBE ACCESS PORT (3/8")WHITE CABINET AND DOORHIGH DENSITY URETHANE FOAM CABINET AND DOOR INSULATIONHFC-FREE REFRIGERANT (R600A) REFRIGERATORNATURAL, HYDROCARBON REFRIGERANT (R290) FREEZERTWO YEAR PARTS AND LABOR WARRANTY, PLUS AN ADDITIONAL THREE YEAR COMPRESSOR PARTS ONLY WARRANTY
Catalog No. NC2468995 Supplier American Biotech Supply Supplier No. PHABTHCRFC1030G
5.2 CU. FT. CAPACITY (REFRIGERATOR) 4 CU. FT. CAPACITY (FREEZER) ONE GLASS SWING DOOR (FRIDGE)ONE SOLID SWING DOOR (FREEZER)3 ADJUSTABLE SHELVES (FREEZER) 3 ADJUSTABLE SHELVES (FRIDGE)BOTTOM FOOTED SHELF TO ALLOW AIR CIRCULATIONBACK WALL EVAPORATOR COVERKEYED DOOR LOCKSMAGNETIC DOOR GASKET FOR POSITIVE SEALRECESSED DOOR HANDLESSTACKING KITLEVELING LEGSLED INTERIOR LIGHTPROBE ACCESS PORT (3/8")WHITE CABINET AND DOORHIGH DENSITY URETHANE FOAM CABINET AND DOOR INSULATIONHFC-FREE REFRIGERANT (R600A) REFRIGERATORNATURAL, HYDROCARBON REFRIGERANT (R290) FREEZERTWO YEAR PARTS AND LABOR WARRANTY, PLUS AN ADDITIONAL THREE YEAR COMPRESSOR PARTS ONLY WARRANTY