Learn More
MINERVA BIOLABS INC Minerva Biolabs Mycoplasma Standard, 10CFU, Mycoplasma arginini, Inactivated
Catalog No. NC3998190
Encompass_Preferred
This mycoplasma standard, featuring 10CFU Mycoplasma arginini, is irreversibly inactivated and packaged as 3 vials with 10 CFU/vial. It is characterized by a GC:CFU ratio of less than 10, with GC content quantified by dPCR.
  • 10 CFU irreversibly inactivated mycoplasma
  • GC:CFU ratio <10 (GC content quantified by dPCR)
  • Product-specific test for inhibitory substances according to revised EP 2.6.7 and upcoming USP <77>
  • Can be used as external positive control
  • Available for 10 relevant Mollicutes species
Catalog No. NC3998190 Supplier MINERVA BIOLABS INC Supplier No. 1051001
This mycoplasma standard, featuring 10CFU Mycoplasma arginini, is irreversibly inactivated and packaged as 3 vials with 10 CFU/vial. It is characterized by a GC:CFU ratio of less than 10, with GC content quantified by dPCR.
  • 10 CFU irreversibly inactivated mycoplasma
  • GC:CFU ratio <10 (GC content quantified by dPCR)
  • Product-specific test for inhibitory substances according to revised EP 2.6.7 and upcoming USP <77>
  • Can be used as external positive control
  • Available for 10 relevant Mollicutes species