Learn More
MINERVA BIOLABS INC 10CFU® Mycoplasma fermentans
Catalog No. NC3998191
Encompass_Preferred
10CFU® Mycoplasma Standards are inactivated, lyophilized mycoplasma particles for product- and matrix-specific validation of NAT-based mycoplasma tests.- EP 2.6.7 (12.2), USP <77>, and JP 18 G3 compliant- ≤10 CFU/ml, <10 GC:CFU- Non-infectious, lyophilized
Catalog No. NC3998191 Supplier MINERVA BIOLABS INC Supplier No. 1056001
10CFU® Mycoplasma Standards are inactivated, lyophilized mycoplasma particles for product- and matrix-specific validation of NAT-based mycoplasma tests.- EP 2.6.7 (12.2), USP <77>, and JP 18 G3 compliant- ≤10 CFU/ml, <10 GC:CFU- Non-infectious, lyophilized