Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MINERVA BIOLABS INC MINERVA BIOLABS Mycoplasma Standard, Mycoplasma hyorhinis, Inactivated, Lyophilized, 10CFU

Catalog No. NC3998195
Encompass_Preferred

This Mycoplasma Standard contains inactivated, lyophilized mycoplasma particles designed for product- and matrix-specific validation of NAT-based mycoplasma tests. It contains 3 vials, each with 10 CFU of irreversibly inactivated Mycoplasma hyorhinis.

  • Contains 3 vials, each with 10 CFU of Mycoplasma hyorhinis
  • Inactivated and lyophilized mycoplasma particles
  • Concentration of ≤10 CFU/ml
  • GC:CFU ratio is <10
  • Compliant with EP 2.6.7 (12.2), USP <77>, and JP 18 G3 regulatory requirements
  • Suitable for method validation, test for inhibitory substances, method suitability test, external positive control, and QC release and routine testing
  • Supports PCR, real-time PCR (qPCR), and digital PCR (dPCR/ddPCR™) detection methods
  • Store at 2°C to 8°C

Catalog No. NC3998195 Supplier MINERVA BIOLABS INC Supplier No. 1057001
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Mycoplasma Standard contains inactivated, lyophilized mycoplasma particles designed for product- and matrix-specific validation of NAT-based mycoplasma tests. It contains 3 vials, each with 10 CFU of irreversibly inactivated Mycoplasma hyorhinis.

  • Contains 3 vials, each with 10 CFU of Mycoplasma hyorhinis
  • Inactivated and lyophilized mycoplasma particles
  • Concentration of ≤10 CFU/ml
  • GC:CFU ratio is <10
  • Compliant with EP 2.6.7 (12.2), USP <77>, and JP 18 G3 regulatory requirements
  • Suitable for method validation, test for inhibitory substances, method suitability test, external positive control, and QC release and routine testing
  • Supports PCR, real-time PCR (qPCR), and digital PCR (dPCR/ddPCR™) detection methods
  • Store at 2°C to 8°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.