Learn More
Cayman Chemical 10Chloroestra1 4dIen3 17 10mg
SDP
Catalog No. 502941997
Encompass_Preferred
A sterone ERα agonist; increases reporter gene expression in S. cerevisiae (EC50 = 0.36 nM for the human receptor); an aromatase inhibitor (IC50 = 2.4 µM)
Catalog No. 50-294-1997 Supplier Cayman Chemical Supplier No. 3975810 MG
A sterone ERα agonist; increases reporter gene expression in S. cerevisiae (EC50 = 0.36 nM for the human receptor); an aromatase inhibitor (IC50 = 2.4 µM)