Learn More
Cayman Chemical 10deACtyl7xylosyl PaclIta 10mg
SDP
Catalog No. 502947754
Encompass_Preferred
A microtubule disruptor; induces the disassembly of pig brain and Physarum microtubules in cell-free assays (IC50s = 0.3 and 0.5 µM, respectively); inhibits proliferation of (IC50 = 5 µM), as well as induces cell cycle arrest at the G2/M phase and apoptosis in, PC3 cells
Catalog No. 50-294-7754 Supplier Cayman Chemical Supplier No. 3295510 MG
A microtubule disruptor; induces the disassembly of pig brain and Physarum microtubules in cell-free assays (IC50s = 0.3 and 0.5 µM, respectively); inhibits proliferation of (IC50 = 5 µM), as well as induces cell cycle arrest at the G2/M phase and apoptosis in, PC3 cells