Learn More
Cayman Chemical 10DSmethoxystreptonIgrIn 2.5mg
SDP
Catalog No. 502941707
Encompass_Preferred
An antibiotic; active against a variety of bacteria, including S. aureus, S. faecalis, E. coli, K. pneumoniae, and P. vulgaris (MICs = 0.4, 1.6, 3.1, 3.1, and 0.4 UG/ml, respectively); cytotoxic to HCT116 colon and A2780 ovarian cancer cells (IC50s = 0.004 and 0.001 UG/ml, respectively); cytotoxic to etoposide- and teniposide-resistant HCT116 cells and cisplatin-resistant A2780 cells (IC50s = 0.003, 0.001, and 0.01 UG/ml, respectively); an inhibitor of p21ras farnesylation (IC50 = 21 nM)
Catalog No. 50-294-1707 Supplier Cayman Chemical Supplier No. 280652.5 MG
An antibiotic; active against a variety of bacteria, including S. aureus, S. faecalis, E. coli, K. pneumoniae, and P. vulgaris (MICs = 0.4, 1.6, 3.1, 3.1, and 0.4 UG/ml, respectively); cytotoxic to HCT116 colon and A2780 ovarian cancer cells (IC50s = 0.004 and 0.001 UG/ml, respectively); cytotoxic to etoposide- and teniposide-resistant HCT116 cells and cisplatin-resistant A2780 cells (IC50s = 0.003, 0.001, and 0.01 UG/ml, respectively); an inhibitor of p21ras farnesylation (IC50 = 21 nM)