Learn More
Trajan Scientific 10F-C/T-GT-5/0.47C SYRINGE
SDP
Catalog No. 502530362
Encompass_Preferred
SGE PAL Autosampler Syringes with 50mm Fixed Cone Tip Needle, Description: Gas tight, Volume: 10 uL, Diameter Outer Needle: 0.47 mm, 0.018 in., Needle Gauge: 26 G, Length Needle: 50 mm, 1.96 in., Connection Type: Fixed, Diameter Inner Needle: 0.004 in., 0.11 mm
Catalog No. 50-253-0362 Supplier Trajan Scientific Supplier No. 002977
SGE PAL Autosampler Syringes with 50mm Fixed Cone Tip Needle, Description: Gas tight, Volume: 10 uL, Diameter Outer Needle: 0.47 mm, 0.018 in., Needle Gauge: 26 G, Length Needle: 50 mm, 1.96 in., Connection Type: Fixed, Diameter Inner Needle: 0.004 in., 0.11 mm