Learn More
Trajan Scientific 10F-CTC-5/0.47C
SDP
Catalog No. 502530394
Encompass_Preferred
SGE PAL Autosampler Syringes with 50 mm Fixed Cone Tip Needle, Volume: 10 uL, Connection Type: Fixed, For Use With: CTC, LEAP, CombiPAL, GC PAL, Gas chromatography, Needle Gauge: 26 G, Needle Point Style: Cone, Description: Not gas tight, Diameter Inner Needle: 0.004 in., 0.11 mm, Quantity: 1/Pk.
Catalog No. 50-253-0394 Supplier Trajan Scientific Supplier No. 002700
SGE PAL Autosampler Syringes with 50 mm Fixed Cone Tip Needle, Volume: 10 uL, Connection Type: Fixed, For Use With: CTC, LEAP, CombiPAL, GC PAL, Gas chromatography, Needle Gauge: 26 G, Needle Point Style: Cone, Description: Not gas tight, Diameter Inner Needle: 0.004 in., 0.11 mm, Quantity: 1/Pk.