Learn More
Trajan Scientific 10F-RSH-5.7/0.63C 10UL SYRINGE
SDP
Catalog No. 502530377
Encompass_Preferred
Trajan Autosampler Syringes With Fixed Needle for Thermo Scientific GCs, Diameter Outer Needle: 0.02 in., 0.63 mm, Needle Gauge: 23 G, Length Needle: 2.2 in., 57 mm, Model: 10F-RTC/RSH-5.7/0.63C, Volume: 10 uL, Connection Type: Fixed
Catalog No. 50-253-0377 Supplier Trajan Scientific Supplier No. 002861
Trajan Autosampler Syringes With Fixed Needle for Thermo Scientific GCs, Diameter Outer Needle: 0.02 in., 0.63 mm, Needle Gauge: 23 G, Length Needle: 2.2 in., 57 mm, Model: 10F-RTC/RSH-5.7/0.63C, Volume: 10 uL, Connection Type: Fixed