Learn More
Trajan Scientific 10F-RTC/RSH-GT-5.7/0.47C 10UL
SDP
Catalog No. 502530374
Encompass_Preferred
Trajan Autosampler Syringes With Fixed Needle for Thermo Scientific GCs, Diameter Outer Needle: 0.01 in., 0.47 mm, Needle Gauge: 26 G, Length Needle: 2.2 in., 57 mm, Model: 10F-RTC/RSH-GT-5.7/0.47C, Volume: 10 uL, Connection Type: Fixed
Catalog No. 50-253-0374 Supplier Trajan Scientific Supplier No. 002866
Trajan Autosampler Syringes With Fixed Needle for Thermo Scientific GCs, Diameter Outer Needle: 0.01 in., 0.47 mm, Needle Gauge: 26 G, Length Needle: 2.2 in., 57 mm, Model: 10F-RTC/RSH-GT-5.7/0.47C, Volume: 10 uL, Connection Type: Fixed