Learn More
Trajan Scientific 10F-RTC/RSH-GT-5.7/0.63C 10UL
SDP
Catalog No. 502530376
Encompass_Preferred
Trajan Autosampler Syringes With Fixed Needle for Thermo Scientific GCs, Diameter Outer Needle: 0.02 in., 0.63 mm, Needle Gauge: 23 G, Length Needle: 2.2 in., 57 mm, Model: 10F-RTC/RSH-GT-5.7/0.63C, Volume: 10 uL, Connection Type: Fixed
Catalog No. 50-253-0376 Supplier Trajan Scientific Supplier No. 002862
Trajan Autosampler Syringes With Fixed Needle for Thermo Scientific GCs, Diameter Outer Needle: 0.02 in., 0.63 mm, Needle Gauge: 23 G, Length Needle: 2.2 in., 57 mm, Model: 10F-RTC/RSH-GT-5.7/0.63C, Volume: 10 uL, Connection Type: Fixed