Learn More
Medchemexpress LLC 10H-Phenoxazine, 4,6-bis(diphenylphosphino)- | 261733-18-0 | 98.1% | 551.55 | 250 MG
SDP
Catalog No. 5000451519
Encompass_Preferred
N-Xantphos is a biochemical reagent that serves as a biological material or organic compound for life science research. It is a functionalized analog of the Xantphos ligand and can be utilized as a supporting ligand for Pd-mediated carbonylation.
  • It is a biochemical reagent.
  • It can be used as a biological material.
  • It can be used as an organic compound for life science related research.
  • It is a functionalized analog of the Xantphos ligand.
  • It can be used as supporting ligand for Pd-mediated carbonylation.
Catalog No. 50-004-51519 Supplier Medchemexpress LLC Supplier No. 250MGHYW008888
N-Xantphos is a biochemical reagent that serves as a biological material or organic compound for life science research. It is a functionalized analog of the Xantphos ligand and can be utilized as a supporting ligand for Pd-mediated carbonylation.
  • It is a biochemical reagent.
  • It can be used as a biological material.
  • It can be used as an organic compound for life science related research.
  • It is a functionalized analog of the Xantphos ligand.
  • It can be used as supporting ligand for Pd-mediated carbonylation.