Learn More
Medchemexpress LLC 10H-Thieno[3,4-b]benzodiazepin-10-one, 4,9-dihydro-3-methyl-4-[2-(4-methyl-1-piperazinyl)acetyl]-, hydrochloride (1:2) | 147416-96-4 | 99.6% | 100 MG
SDP
Catalog No. 5000412825
Encompass_Preferred
Telenzepine dihydrochloride is a selective and orally active muscarinic M1 receptor antagonist with a Ki of 0.94 nM. It inhibits gastric acid secretion and has antiulcer effects. It is for research use only.
  • Selective and orally active muscarinic M1 receptor antagonist with a Ki of 0.94 nM.
  • Inhibits gastric acid secretion.
  • Has antiulcer effects.
Catalog No. 50-004-12825 Supplier Medchemexpress LLC Supplier No. HYB1789A100MG
Telenzepine dihydrochloride is a selective and orally active muscarinic M1 receptor antagonist with a Ki of 0.94 nM. It inhibits gastric acid secretion and has antiulcer effects. It is for research use only.
  • Selective and orally active muscarinic M1 receptor antagonist with a Ki of 0.94 nM.
  • Inhibits gastric acid secretion.
  • Has antiulcer effects.