Learn More
Cayman Chemical 10hydroxy12 ZOctadece 100ug
SDP
Catalog No. 502917960
Encompass_Preferred
An internal standard for the quantification of (±)-10-hydroxy-12(Z)-octadecenoic acid by GC- or LC-MS
Catalog No. 50-291-7960 Supplier Cayman Chemical Supplier No. 37282100 UG
An internal standard for the quantification of (±)-10-hydroxy-12(Z)-octadecenoic acid by GC- or LC-MS