Learn More
Cole-Parmer Type-K, E x tra-Long PFA Coated Probe, Mini-Connector, 10" L. .145" Dia, Grounded
Catalog No. NC9273713 Shop All Cole-Parmer Products
Encompass_Preferred
General-Purpose probes with PTFE-coating are designed for use with corrosive chemicals and strong acids. Response Time: 2 seconds or greater. ABS handles provide a light weight design for ease of use along with durability. Probe handles feature a strain relief spring that protects the cable connection against damage due to repeated flexing and tugging.
Catalog No. NC9273713 Supplier Cole-Parmer Supplier No. 9381202
General-Purpose probes with PTFE-coating are designed for use with corrosive chemicals and strong acids. Response Time: 2 seconds or greater. ABS handles provide a light weight design for ease of use along with durability. Probe handles feature a strain relief spring that protects the cable connection against damage due to repeated flexing and tugging.