Learn More
Chemglass Life Sciences LabJack, Anodized Aluminum, 25 x 20cm (10 x 7.8") Top Plate, 32cm (12.5") Vertical Lift
SDP
Catalog No. 503903
Encompass_Preferred
LabJack, Anodized Aluminum, 25 x 20cm (10 x 7.8") Top Plate, 32cm (12.5") Vertical Lift. Support Jack provide for stable height adjustment and support for items in the lab such as flasks, baths or hot plate stirrers. Top and bottom plates are constructed of anodized aluminum while the support mechanism is made of chrome plated steel. Plates have a threaded hole for the optional vertical support rod which is made of stainless steel and is 11mm OD x 17¿ tall. Max Load 250lbs.
Catalog No. 50-390-3 Supplier Chemglass Life Sciences Supplier No. CG305414
LabJack, Anodized Aluminum, 25 x 20cm (10 x 7.8") Top Plate, 32cm (12.5") Vertical Lift. Support Jack provide for stable height adjustment and support for items in the lab such as flasks, baths or hot plate stirrers. Top and bottom plates are constructed of anodized aluminum while the support mechanism is made of chrome plated steel. Plates have a threaded hole for the optional vertical support rod which is made of stainless steel and is 11mm OD x 17¿ tall. Max Load 250lbs.