Learn More
Chemglass Life Sciences LabJack, Stainless Steel, 25 x 25cm (10 x 10") Top Plate, 33cm (13in) Vertical Lift
SDP
Catalog No. 504879
Encompass_Preferred
LabJack, Stainless Steel, 25 x 25cm (10 x 10") Top Plate, 33cm (13in) Vertical Lift. Support Jack provide for stable height adjustment and support for items in the lab such as flasks, baths or hot plate stirrers. Top and bottom plates are constructed of stainless steel while the support mechanism is made of chrome plated steel.
Catalog No. 50-487-9 Supplier Chemglass Life Sciences Supplier No. CG305625
LabJack, Stainless Steel, 25 x 25cm (10 x 10") Top Plate, 33cm (13in) Vertical Lift. Support Jack provide for stable height adjustment and support for items in the lab such as flasks, baths or hot plate stirrers. Top and bottom plates are constructed of stainless steel while the support mechanism is made of chrome plated steel.