Learn More
Abcam 10kD Spin Column, 25UNIT
Supplier:  Abcam AB9334925UNIT
Encompass_Preferred
10kD spin column is a disposable ultrafiltration device designed for the concentration or deproteinization of biological samples. - Cited in over 25 publications - Compatible with swing bucket or fixed-angle rotors at up to 10,000 x g - Low internal surface and membrane area for optimal recovery from dilute solutions - Sample capacity – maximum initial sample volume of 500 µL, with a dead stop volume of 20-25 µL - Cell lysate compatible – recommended to centrifuge lysates prior to use to avoid membrane blockage
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-4787