Learn More
Worthington Biochemical Corporation Deoxyribonuclease I, Ribonuclease&Protease Free, 10ku
SDP
Catalog No. NC9082558
Encompass
Molecular Biology Grade. Chromatographically purified to remove RNase and protease. Lyophilized in vial containing glycine and calcium as stabilizer. Store at 2-8°C. Protect from moisture. Bovine pancreatic deoxyribonuclease is an endonuclease which splits phosphodiester linkages, preferentially adjacent to a pyrimidine nucleotide yielding polynucleotides with free hydroxyl group at the 3' position and phosphate group at the 5' position. The average chain length of a limit digest is a tetranucleotide.
Catalog No. NC9082558 Supplier Worthington Biochemical Corporation Supplier No. LS006333
Molecular Biology Grade. Chromatographically purified to remove RNase and protease. Lyophilized in vial containing glycine and calcium as stabilizer. Store at 2-8°C. Protect from moisture. Bovine pancreatic deoxyribonuclease is an endonuclease which splits phosphodiester linkages, preferentially adjacent to a pyrimidine nucleotide yielding polynucleotides with free hydroxyl group at the 3' position and phosphate group at the 5' position. The average chain length of a limit digest is a tetranucleotide.