Learn More
Ace Glass, Inc. Mantle, 10L, for 193.8mm diameter cylindrical flask, 193.8mm ID x 56.4mm height, 180w-115v, CSA rated
SDP
Catalog No. 50983199
Encompass
10L aluminum housed, table-model mantle for 193.8mm diameter cylindrical flask. 193.8mm I.D. x 56.4mm height. Maximum internal operating temperature is 450?C. 180w-115v, and is cULus rated. Power cord included.
Catalog No. 50-983-199 Supplier Ace Glass, Inc. Supplier No. 1205350
10L aluminum housed, table-model mantle for 193.8mm diameter cylindrical flask. 193.8mm I.D. x 56.4mm height. Maximum internal operating temperature is 450?C. 180w-115v, and is cULus rated. Power cord included.