Learn More
Molecular Devices Molecular Devices Waste Reservoir, 10 L, AquaMax compatible
Catalog No. 5000014372
Encompass_Preferred
Designed for Molecular Devices AquaMax microplate washers, this 10 L waste reservoir provides a large-capacity, replaceable container for spent wash solutions. It reduces maintenance frequency and simplifies fluid handling during routine microplate washing operations.
  • Capacity 10 L.
  • Compatible with 2000 and 4000 series, SW400, and DW4 microplate washers.
  • Reduces maintenance frequency by increasing waste capacity.
  • Simple installation and replacement for routine maintenance.
  • Collects and contains spent wash solutions for simplified disposal.
Catalog No. 50-000-14372 Supplier Molecular Devices Supplier No. 90000761
Designed for Molecular Devices AquaMax microplate washers, this 10 L waste reservoir provides a large-capacity, replaceable container for spent wash solutions. It reduces maintenance frequency and simplifies fluid handling during routine microplate washing operations.
  • Capacity 10 L.
  • Compatible with 2000 and 4000 series, SW400, and DW4 microplate washers.
  • Reduces maintenance frequency by increasing waste capacity.
  • Simple installation and replacement for routine maintenance.
  • Collects and contains spent wash solutions for simplified disposal.