Learn More
Sanisure Inc PharmaTainer™ Bottle - Polycarbonate
Catalog No. NC3254383
Encompass_Preferred
Our PharmaTainer™ Bottle Solutions range from 10 mL to 20 L single-use bottles in PET or PC. This sterile solution can be used for liquid transfer, storage, and transporting vaccines, biopharmaceuticals, culture media, or other biotech materials.
Catalog No. NC3254383 Supplier Sanisure Inc Supplier No. BF25501S
Our PharmaTainer™ Bottle Solutions range from 10 mL to 20 L single-use bottles in PET or PC. This sterile solution can be used for liquid transfer, storage, and transporting vaccines, biopharmaceuticals, culture media, or other biotech materials.