Learn More
Chemglass Life Sciences 10L Process Reactor, Complete, Air Motor, 67.0" OAH with Motor, 15.3" Clearance Below Valve
SDP
Catalog No. CG1964610
Encompass_Preferred
10L Process Reactor, Complete, Air Motor, 67.0" OAH with Motor, 15.3" Clearance Below Valve. Features:¿ Open Frame Design Provides Excellent Accessibility¿ Support Frame with Oversized Rugged Stainless Steel Uprights and Heavy Duty PTFE Coated Aluminum Fittings¿ Perfluoro O-Rings On 60mm and 200mm Flanges¿ Stainless Steel Inlet/Outlet Adapters Available In M16 x 1 and M30 x 1.5 Allow Connection To Cooling/Heating Circulator¿ Lower Support Plate, and PTFE Ring, Have a "V" Shaped Opening Allowing An Unobstructed View Of Bottom Valve¿ Zero Dead Space Detachable Drain Valve Has Knob with Laser Etched Directional Arrows For Open and Close¿ Electric or Air Motor. Optional XP Motor Available¿ 5-Neck Lid with Large Powder Addition Funnel¿ Digital Temperature Monitor¿ Electric Motor Has Separate Digital Controller¿ Reactor Jacket Graduated On Both Sides¿ Support Bushings For Optional Safety Shield¿ Heavy Duty Lockable Casters
Catalog No. CG1964610 Supplier Chemglass Life Sciences Supplier No. CG1964610
Add to Cart
Add to Cart
Request a Quote
10L Process Reactor, Complete, Air Motor, 67.0" OAH with Motor, 15.3" Clearance Below Valve. Features:¿ Open Frame Design Provides Excellent Accessibility¿ Support Frame with Oversized Rugged Stainless Steel Uprights and Heavy Duty PTFE Coated Aluminum Fittings¿ Perfluoro O-Rings On 60mm and 200mm Flanges¿ Stainless Steel Inlet/Outlet Adapters Available In M16 x 1 and M30 x 1.5 Allow Connection To Cooling/Heating Circulator¿ Lower Support Plate, and PTFE Ring, Have a "V" Shaped Opening Allowing An Unobstructed View Of Bottom Valve¿ Zero Dead Space Detachable Drain Valve Has Knob with Laser Etched Directional Arrows For Open and Close¿ Electric or Air Motor. Optional XP Motor Available¿ 5-Neck Lid with Large Powder Addition Funnel¿ Digital Temperature Monitor¿ Electric Motor Has Separate Digital Controller¿ Reactor Jacket Graduated On Both Sides¿ Support Bushings For Optional Safety Shield¿ Heavy Duty Lockable Casters