Learn More
Chemglass Life Sciences 10L Process Reactor, Complete, Electric Motor, 70.1" OAH with Motor, 15.3" Clearance Below Valve
SDP
Catalog No. CG1967610
Encompass_Preferred
10L Process Reactor, Complete, Electric Motor, 70.1" OAH with Motor, 15.3" Clearance Below Valve. ¿ New & Improved Open Frame Design Provides Excellent Accessibility¿ 6-Neck Lid With Large Powder Addition Port¿ Perfluoro O-Rings On 60mm and 200mm Flanges¿ Improved Support Clamp Auto-Centers Reactor and Lid Allowing Quick Installation/Removal Of Reactor¿ #304 S.S. Cooling Coil PTFE Coated¿ Lower Support Plate, and PTFE Ring, Have a "V" Shaped Opening Allowing An Unobstructed View Of Bottom Valve¿ Zero Dead Space Detachable Drain Valve Has Knob with Laser Etched Directional Arrows For Open and Close¿ Electric or Air Motor. Optional XP Motor Available¿ Electric Motor Has Separate Digital Controller¿ Digital Temperature Controller¿ Support Frame with Oversized Rugged S.S.Uprights and Heavy Duty PTFE Coated Aluminum Fittings¿ Removable S.S. Pipe Used To Support Side Shelf and Manifold System¿ Support Bushings For Optional Safety Shield¿ Heavy Duty Lockable Casters
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. CG1967610 Supplier Chemglass Life Sciences Supplier No. CG1967610
Add to Cart
Add to Cart
Request a Quote
10L Process Reactor, Complete, Electric Motor, 70.1" OAH with Motor, 15.3" Clearance Below Valve. ¿ New & Improved Open Frame Design Provides Excellent Accessibility¿ 6-Neck Lid With Large Powder Addition Port¿ Perfluoro O-Rings On 60mm and 200mm Flanges¿ Improved Support Clamp Auto-Centers Reactor and Lid Allowing Quick Installation/Removal Of Reactor¿ #304 S.S. Cooling Coil PTFE Coated¿ Lower Support Plate, and PTFE Ring, Have a "V" Shaped Opening Allowing An Unobstructed View Of Bottom Valve¿ Zero Dead Space Detachable Drain Valve Has Knob with Laser Etched Directional Arrows For Open and Close¿ Electric or Air Motor. Optional XP Motor Available¿ Electric Motor Has Separate Digital Controller¿ Digital Temperature Controller¿ Support Frame with Oversized Rugged S.S.Uprights and Heavy Duty PTFE Coated Aluminum Fittings¿ Removable S.S. Pipe Used To Support Side Shelf and Manifold System¿ Support Bushings For Optional Safety Shield¿ Heavy Duty Lockable Casters
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.