Learn More
Alkali Scientific 10L H2010 Shaking Incubator Wi
SDP
Catalog No. 503029593
Encompass_Preferred
The Alkali Scientific10L H2010 Shaking Incubator With Touch Screen Programmable Interface is a heavy-duty performer designed for heavy workloads and continuous use Its outstanding temperature accuracy and uniformity are supported by proprietary heat distribution technology that ensures that temperatures are precisely maintained throughout the chamber Inconsistencies due to stratification are reduced to negligible levels and the selected temperature is precisely maintained within 0 25% even in the corners The powerful brushless induction drive is designed for years of trouble-free service even under continuous use conditions with heavy loads Noise and vibration are minimized by an advanced computer designed motor isolation system Like other Incu-Shaker models the 10L is built to last A standard rubber mat platform is included and is ideal for use with tissue culture flasks Petri dishes staining trays and other flat vessels The convenient MAGic Clampplatformis also available for u
Catalog No. 50-302-9593 Supplier Alkali Scientific Supplier No. RS7090
Add to Cart
Add to Cart
Request a Quote
The Alkali Scientific10L H2010 Shaking Incubator With Touch Screen Programmable Interface is a heavy-duty performer designed for heavy workloads and continuous use Its outstanding temperature accuracy and uniformity are supported by proprietary heat distribution technology that ensures that temperatures are precisely maintained throughout the chamber Inconsistencies due to stratification are reduced to negligible levels and the selected temperature is precisely maintained within 0 25% even in the corners The powerful brushless induction drive is designed for years of trouble-free service even under continuous use conditions with heavy loads Noise and vibration are minimized by an advanced computer designed motor isolation system Like other Incu-Shaker models the 10L is built to last A standard rubber mat platform is included and is ideal for use with tissue culture flasks Petri dishes staining trays and other flat vessels The convenient MAGic Clampplatformis also available for u