Learn More
Ace Glass, Inc. Beaker, 10L, heavy wall, 225mm OD x 340mm height, not graduated, Duran;
SDP
Catalog No. 509557018
Encompass
10L Duran heavy wall beaker with spout. Beaker is not graduated. Dimensions: 225mm O.D. x 340mm Height.
Catalog No. 50-955-7018 Supplier Ace Glass, Inc. Supplier No. 533233
10L Duran heavy wall beaker with spout. Beaker is not graduated. Dimensions: 225mm O.D. x 340mm Height.