Learn More
Foxx Life Sciences 10L PP Carboy, Sterile
GREENER_CHOICE
Catalog No. 5000846717
Encompass_Preferred
EZclear 10L Polypropylene PP Carboy is a translucent container that allows easy visual inspection of its contents. The single use 10L PP carboy maintains durability, chemical resistance, and autoclavability of standard polypropylene carboys. The 10L PP Wide mouth carboys features include large top handles, easy-to-read metric graduations (±5% certified accuracy). The 10L sterile PP Carboys manufactured from USP Class VI, polypropylene resin, offering good chemical resistance and thermal stability. 10L Autoclavable PP carboys have a recommended sterilization cycle of 121 °C at 15 psi for 20 minutes.
Features:
VersaCap technology enables flexible, interchangeable connector options
Transparency with approximately 30% haze
Leak-proof cap design
Double bagged sterile carboy
10L PP Round Carboys ideal for mixing
Made in Class 7 Cleanroom environment
Catalog No. 50-008-46717 Supplier Foxx Life Sciences Supplier No. 515EG52FLS
EZclear 10L Polypropylene PP Carboy is a translucent container that allows easy visual inspection of its contents. The single use 10L PP carboy maintains durability, chemical resistance, and autoclavability of standard polypropylene carboys. The 10L PP Wide mouth carboys features include large top handles, easy-to-read metric graduations (±5% certified accuracy). The 10L sterile PP Carboys manufactured from USP Class VI, polypropylene resin, offering good chemical resistance and thermal stability. 10L Autoclavable PP carboys have a recommended sterilization cycle of 121 °C at 15 psi for 20 minutes.
Features:
VersaCap technology enables flexible, interchangeable connector options
Transparency with approximately 30% haze
Leak-proof cap design
Double bagged sterile carboy
10L PP Round Carboys ideal for mixing
Made in Class 7 Cleanroom environment