Learn More
Chemglass Life Sciences 10L UnJacketed Cylindrical Process Filter Reactor System Complete, Air Motor
SDP
Catalog No. 501531302
Encompass_Preferred
10L UnJacketed Cylindrical Process Filter Reactor System Complete, Air Motor. Chemglass's unjacketed filter reactors can be used for single or multi-step reactions and filtrations in the same vessel, production and process scale peptide synthesis. Filter base eliminates material transfers and significantly reduces losses. Complete reactor system on Chemglass's open air, autocenter stand. Please Note: Filter Reactors are for Vacuum/Gravity Filtration Use Only
Catalog No. 50-153-1302 Supplier Chemglass Life Sciences Supplier No. CG1958F811
Add to Cart
Add to Cart
Request a Quote
10L UnJacketed Cylindrical Process Filter Reactor System Complete, Air Motor. Chemglass's unjacketed filter reactors can be used for single or multi-step reactions and filtrations in the same vessel, production and process scale peptide synthesis. Filter base eliminates material transfers and significantly reduces losses. Complete reactor system on Chemglass's open air, autocenter stand. Please Note: Filter Reactors are for Vacuum/Gravity Filtration Use Only