Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Foxx Life Sciences PUREGRIP Rhino Safety Coated Glass Carboy, Clear, 10L, with Graduation Marks and 83B VersaCap, 1/EA

Supplier:  Foxx Life Sciences 48RC800FLS

Encompass_Preferred

PUREGRIP Rhino Bottles & Carboys excel in mixing, storing, and transporting media, buffers, chemicals, and solvents while enhancing lab safety. The Rhino Safety coating secures glass fragments if bottles break, minimizing risks, while its non-slip grip improves wet handling for a safer, more efficient lab. Made from durable 3.3 Borosilicate Glass and enabled with graduation marks to ensure precise measurements.
These products enhance lab safety through drip-free dispensing, minimizing spills, limiting chemical exposure, and enabling quick cleanup—an ideal choice for safety-focused labs.

Features

Shatter Resistant Coating
Non-Slip Exterior
Clear and Amber Options Available in sizes from 100mL to 20L
Thermal & Chemical Resistance: Withstands 121°C autoclaving and -20°C refrigeration
USP Class VI Certified: Includes an animal origin free polypropylene VersaCap with Secure Grasp technology for easy handling.

Catalog No. 50-297-8692


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.