Learn More
Chemglass Life Sciences Chemglass CG-1924-R-10L 10 L Reaction Vessel, Unjacketed, Graduated, 200 mm Flange, 0-25 mm Spring-Loaded Drain Valve
SDP
Supplier:  Chemglass Life Sciences CG1924R10L
Encompass_Preferred
Chemglass 10 L unjacketed reaction vessel for process-scale laboratory reactions. It features graduated markings for approximate volume, a 200 mm flange designed for greased seals with compatible lids, and a spring-loaded bottom drain for controlled discharge across a wide temperature range.
  • 10 L capacity.
  • Unjacketed cylindrical design.
  • Graduated markings on both sides for approximate volume.
  • 200 mm flange with groove in sealing surface for greased seal compatibility.
  • 0-25 mm spring-loaded drain valve rated for -60° C to 230° C with perfluoro Class VI O-rings.
  • Drain valve sidearm with 3/4 in Swagelok connection.
  • Flange seals flush with bottom to minimize dead space.
  • Serial numbered for traceability.
Catalog No. NC3822340