Learn More
Alkali Scientific 1000µl Refill Pipet Tips,1mL, (8 Racks of 96), 768/cs
SDP
Catalog No. 502335945
Encompass_Preferred
PurePoint™™ Reload Pipette Tip Box are the economical choice when it comes to liquid handling. Simply replace your empty racks with these convenient wafers. These refill pipet tips fit into Alkali Scientific's PurePoint™ Pipet Tip boxes and are universal fit to standard pipettors. The tips are ultra-clear and graduated, and they are packaged Rnase/Dnase and pyrogen free so you can be confident your sample results will not contain unwanted contaminates. Refill pipet tips are a more environmentally friendly liquid handling system. Refills use less plastic by utilizing the tip boxes from PurePoint™ pipet tips and filter pipet tips allowing for less waste. They are made from 100% pure polypropylene plastic and are low-retention leaving less fluid in the tip following ejection. Recommended autoclave procedure: 120° for 12 minutes @ 15 PSI.
Catalog No. 50-233-5945 Supplier Alkali Scientific Supplier No. RT1000
PurePoint™™ Reload Pipette Tip Box are the economical choice when it comes to liquid handling. Simply replace your empty racks with these convenient wafers. These refill pipet tips fit into Alkali Scientific's PurePoint™ Pipet Tip boxes and are universal fit to standard pipettors. The tips are ultra-clear and graduated, and they are packaged Rnase/Dnase and pyrogen free so you can be confident your sample results will not contain unwanted contaminates. Refill pipet tips are a more environmentally friendly liquid handling system. Refills use less plastic by utilizing the tip boxes from PurePoint™ pipet tips and filter pipet tips allowing for less waste. They are made from 100% pure polypropylene plastic and are low-retention leaving less fluid in the tip following ejection. Recommended autoclave procedure: 120° for 12 minutes @ 15 PSI.