Learn More
Ace Glass, Inc. 10L StreamFlow Bottle with Hose Connection includes expanded PTFE membrane vented cap GL-45 thread hose connection for 5/8 id tubing
SDP
Catalog No. 5000810390
Encompass_Preferred
10L StreamFlow Bottle with Hose Connection includes expanded PTFE membrane vented cap GL-45 thread hose connection for 5/8 id tubing
Catalog No. 50-008-10390 Supplier Ace Glass, Inc. Supplier No. 538908
10L StreamFlow Bottle with Hose Connection includes expanded PTFE membrane vented cap GL-45 thread hose connection for 5/8 id tubing