Learn More
Beckman Coulter CytoFLEX, Research Flow Cytometer
Catalog No. NC1743673
Encompass
The Beckman Coulter CytoFLEX is a benchtop flow cytometer with 13 fluorescent channels, 3 lasers (405nm, 488nm, 638nm), and analyzes up to 30,000 events/sec. It's designed for high sensitivity and resolution, offering applications like immunophenotyping and apoptosis. Compact and user-friendly, it includes intuitive software for diverse research needs.
Catalog No. NC1743673 Supplier Beckman Coulter Supplier No. B86549
The Beckman Coulter CytoFLEX is a benchtop flow cytometer with 13 fluorescent channels, 3 lasers (405nm, 488nm, 638nm), and analyzes up to 30,000 events/sec. It's designed for high sensitivity and resolution, offering applications like immunophenotyping and apoptosis. Compact and user-friendly, it includes intuitive software for diverse research needs.