Learn More
Cayman Chemical 10-methyl-9-(phenoxycarbonyl) Acridinium (trifluoromethylsulfonate), Purity: ≥95%, CAS: 161006-14-0, 50 mg
SDP
Catalog No. 502933056
Encompass_Preferred
An acridinium ester that produces fluorescent 10-methyl-9-acridone upon oxidation with hydrogen peroxide, persulfates, and other oxidants in alkaline conditions; can be used in chemiluminescence assays, enzyme, antigen, antibody, and hormone immunoassays, and assays to detect oxidants in environmental, biological, and pharmaceutical samples
Catalog No. 50-293-3056 Supplier Cayman Chemical Supplier No. 2588550 MG
An acridinium ester that produces fluorescent 10-methyl-9-acridone upon oxidation with hydrogen peroxide, persulfates, and other oxidants in alkaline conditions; can be used in chemiluminescence assays, enzyme, antigen, antibody, and hormone immunoassays, and assays to detect oxidants in environmental, biological, and pharmaceutical samples