Learn More
Selleck Chemical LLC Colforsin (Forskolin) 10mg 66575-29-9 Coleonol
SDP
Catalog No. NC1216734
Encompass_Preferred
Colforsin (Forskolin, Coleonol) is a ubiquitous activator of eukaryotic adenylyl cyclase (AC) in a wide variety of cell types, commonly used to raise levels of cAMP in the study and research of cell physiology. Forskolin also activates PXR and FXR activity. Forskolin stimulates autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1216734 Supplier Selleck Chemical LLC Supplier No. S244910MG
Colforsin (Forskolin, Coleonol) is a ubiquitous activator of eukaryotic adenylyl cyclase (AC) in a wide variety of cell types, commonly used to raise levels of cAMP in the study and research of cell physiology. Forskolin also activates PXR and FXR activity. Forskolin stimulates autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.