Learn More
Cayman Chemical MK-571, 115104-28-4, 10 mg
SDP
Supplier:  Cayman Chemical 1002910
Encompass_Preferred
Molecular Formula C26H27ClN2O3S2, Purity ≥95%, Formula Weight 515.1, Synonyms: L-660,711; A selective, orally active CysLT1 receptor antagonist; blocks the binding of LTD4 to human and guinea pig lung membranes with Ki values of 0.22 nM and 2.1 nM, respectively.
Catalog No. NC0670204