Learn More
Invivogen EndoFit Ovalbumin, Chicken Egg Albumin, for In Vivo Use, 10 mg
SDP
Supplier:  Invivogen VACPOVA
Encompass
Ovalbumin (also known as albumin or OVA) is a key reference protein for immunization. Ovalbumin is the most abundant protein in chicken egg whites. Ovalbumin is a glycoprotein that is sufficiently large and complex to be mildly immunogenic. Consequently, ovalbumin is widely used as an antigen for immunization experiments [1-3]. Furthermore, ovalbumin can be used as a carrier protein for conjugation to haptens [4] and other antigens to make them more immunogenic.
Catalog No. NC0456409