Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 10mg, Porphobilinogen, >97%, [487-90-1], P226 244.24 C10H14N2O4 x H2O

Catalog No. 5039001
Encompass_Preferred

Compound: Porphobilinogen

CAS#: 487-90-1

MDL#: MFCD00005224

Background: Porphobilinogen is a pyrrole-based natural product that serves as the starting material for the biosynthesis of porphyrins such as heme and chlorophyll. It is a main indicator of altered metabolism, with elevated levels indicative of lead poisoning, and serves as a biomarker for porphyria disease.

Catalog No. 50-390-01 Supplier Frontier Specialty Chemicals Supplier No. P22610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Porphobilinogen

CAS#: 487-90-1

MDL#: MFCD00005224

Background: Porphobilinogen is a pyrrole-based natural product that serves as the starting material for the biosynthesis of porphyrins such as heme and chlorophyll. It is a main indicator of altered metabolism, with elevated levels indicative of lead poisoning, and serves as a biomarker for porphyria disease.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.