Learn More
Ace Glass, Inc. 10mL graduated cylindrical funnel, 14/20 top, 14/20 bottom, metering valve stopcock plug
SDP
Catalog No. 50976957
Encompass_Preferred
10mL graduated cylindrical funnel with 14/20 top and bottom joints, a drip stem, and double scale graduations that are calibrated from the stopcock. Drip stem does not extend beyond the joint, thereby eliminating a possible source of breakage. Funnel has a metering valve stopcock plug.
Catalog No. 50-976-957 Supplier Ace Glass, Inc. Supplier No. 949904
10mL graduated cylindrical funnel with 14/20 top and bottom joints, a drip stem, and double scale graduations that are calibrated from the stopcock. Drip stem does not extend beyond the joint, thereby eliminating a possible source of breakage. Funnel has a metering valve stopcock plug.