Learn More
Thomas Scientific 24-Well Polypropylene Storage/Reaction Microplate, 10mL/square well, round bottoms, 44mm height, 25/cs
SDP
Catalog No. NC1096282
Encompass
Agilent supplies collection and storage assay microplates for a wide range of reaction chemistries, biological assays, low storage temperatures, and filtrate collection applications. These microplates are molded with medical-grade, chemical-resistant polymers and are available in 24-, 48-, 96-, or 384-well configurations of variable volumes. Agilent also supplies mat seals and lids for these microplates.
Catalog No. NC1096282 Supplier Thomas Scientific Supplier No. 1149Q53
Agilent supplies collection and storage assay microplates for a wide range of reaction chemistries, biological assays, low storage temperatures, and filtrate collection applications. These microplates are molded with medical-grade, chemical-resistant polymers and are available in 24-, 48-, 96-, or 384-well configurations of variable volumes. Agilent also supplies mat seals and lids for these microplates.