Learn More
Biophoretics Inc SERVALYT 9-11 Carrier Ampholytes, 10 mL, 40% aqueous concentrate, pH range 9-11, analytical grade, IEF, cIEF, free-flow electrophoresis
Catalog No. NC0737821
Encompass
SERVALYT 9-11 Carrier Ampholytes, 10 mL, 40% aqueous concentrate, pH range 9-11, analytical grade, IEF, cIEF, free-flow electrophoresis
Catalog No. NC0737821 Supplier Biophoretics Inc Supplier No. 42909.01
SERVALYT 9-11 Carrier Ampholytes, 10 mL, 40% aqueous concentrate, pH range 9-11, analytical grade, IEF, cIEF, free-flow electrophoresis