Learn More
Analytical Sales & Services 10mL Amber Glass Crimp Style Head Space Vial
Catalog No. 5000804592
Encompass
Advantage™ 20mm, 10mL Volume, 23 x 46mm Amber Glass Crimp Style Head Space Vial with Beveled Top and Round Bottom - for CTC Autsamplers
Catalog No. 50-008-04592 Supplier Analytical Sales & Services Supplier No. 611823
Advantage™ 20mm, 10mL Volume, 23 x 46mm Amber Glass Crimp Style Head Space Vial with Beveled Top and Round Bottom - for CTC Autsamplers