Learn More
Alkali Scientific 10mL Amber Glass Crimp Top Hea
SDP
Catalog No. 503028807
Encompass_Preferred
Designed for precise chromatography analysis ASI10 mL Amber Glass Round Flat-Bottom Crimp Top Headspace Vials provide an affordable high-quality solution compatible with all headspace GC instruments Manufactured from amber Type I borosilicate glass these vials offer exceptional thermal and chemical resistance while ensuring optimal protection for light-sensitive samples The round flat-bottom design enhances heating efficiency and ensures compatibility with a wide range of headspace autosamplers Key Features Amber Borosilicate Glass Protects light-sensitive samples from UV exposure while maintaining high chemical resistance and durability Beveled Finish Reduces the sealing surface between the crimp seal and the glass for a more secure and consistent closure Standard 20 mm Crimp Top Ensures compatibility with a wide range of crimp seals and autosamplers Optimal Size for Headspace Analysis 22 5 46 mm dimensions allow precise sample containment Round Flat-Bottom Design Ideal f
Catalog No. 50-302-8807 Supplier Alkali Scientific Supplier No. GC010FBAMB
Designed for precise chromatography analysis ASI10 mL Amber Glass Round Flat-Bottom Crimp Top Headspace Vials provide an affordable high-quality solution compatible with all headspace GC instruments Manufactured from amber Type I borosilicate glass these vials offer exceptional thermal and chemical resistance while ensuring optimal protection for light-sensitive samples The round flat-bottom design enhances heating efficiency and ensures compatibility with a wide range of headspace autosamplers Key Features Amber Borosilicate Glass Protects light-sensitive samples from UV exposure while maintaining high chemical resistance and durability Beveled Finish Reduces the sealing surface between the crimp seal and the glass for a more secure and consistent closure Standard 20 mm Crimp Top Ensures compatibility with a wide range of crimp seals and autosamplers Optimal Size for Headspace Analysis 22 5 46 mm dimensions allow precise sample containment Round Flat-Bottom Design Ideal f