Learn More
Gerstel 10ML AMBER VIAL W/SCREW NECK
SDP
Catalog No. NC0575021
Encompass
10 mL amber screw cap vials for MPS (rounded bottom for transport), Package of 100 Requires magnetic screw caps with septa 093640-092-00 (PolyMag) or 093640-040-00 (metal)
Catalog No. NC0575021 Supplier Gerstel Supplier No. 09364003900
10 mL amber screw cap vials for MPS (rounded bottom for transport), Package of 100 Requires magnetic screw caps with septa 093640-092-00 (PolyMag) or 093640-040-00 (metal)