Learn More
CELLTREAT Scientific Products 10mL Aspirating Pipet, Short, Individually Wrapped, Sterile
SDP
Catalog No. NC3796188
Encompass_Preferred
Ideal for aspiration of media and sample waste removal without needing to measure the volumeAspirating pipets are supplied without graduations or plugsCompact design of short aspirating pipet provides ergonomic benefit and reduces storage spaceShorter length helps prevent touching pipet tip to surfaces in the hood"
Catalog No. NC3796188 Supplier CELLTREAT Scientific Products Supplier No. 229237
Ideal for aspiration of media and sample waste removal without needing to measure the volumeAspirating pipets are supplied without graduations or plugsCompact design of short aspirating pipet provides ergonomic benefit and reduces storage spaceShorter length helps prevent touching pipet tip to surfaces in the hood"