Learn More
S2 Media 10mL Butterfield's Phosphate Buffer (Phosphate Buffer, pH 7.2), 15mL Conical Tube, 50/box
Catalog No. 502995755
Encompass_Preferred
Butterfield's Phosphate Buffer (BPB), pH 7.2, is utilized as a diluent in the preparation of sample dilutions and microbiological suspensions in laboratory processes. • Product meets AOAC, APHA, ASM, BAM, SM-Dairy, SMWW, Harmonized USP/EP/JP, and USDA reference methods and performance criteria.• Butterfield's Phosphate Buffer should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Also known as Phosphate Buffer, pH 7.2, and Butterfield's Phosphate Buffered Dilution Water.
This item is not returnable. View return policy
Catalog No. 50-299-5755 Supplier S2 Media Supplier No. 5196
Butterfield's Phosphate Buffer (BPB), pH 7.2, is utilized as a diluent in the preparation of sample dilutions and microbiological suspensions in laboratory processes. • Product meets AOAC, APHA, ASM, BAM, SM-Dairy, SMWW, Harmonized USP/EP/JP, and USDA reference methods and performance criteria.• Butterfield's Phosphate Buffer should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Also known as Phosphate Buffer, pH 7.2, and Butterfield's Phosphate Buffered Dilution Water.