Learn More
CELLTREAT Scientific Products 10mL Centrifuge Tube, Oak Ridge Style, Round Bottom, PP, Screw Cap, Non-sterile, 12/case
SDP
Catalog No. NC3433461
Encompass_Preferred
"High speed tubes feature a round bottom and uniform wall thicknessTubes are economical and leak-proof for refrigerated or standard centrifugationTube should be filled 80% of total capacity to obtain maximum results"
Catalog No. NC3433461 Supplier CELLTREAT Scientific Products Supplier No. 229460
"High speed tubes feature a round bottom and uniform wall thicknessTubes are economical and leak-proof for refrigerated or standard centrifugationTube should be filled 80% of total capacity to obtain maximum results"