Learn More
MTC Bio 10ml PP (16x104mm), flat screw cap, 50 tubes per sterile bag
Catalog No. NC2017618
Encompass_Preferred
Conical 10ml, 15ml, and 50ml centrifuge tubes are made of premium polypropylene for clarity and centrifugal strength. Tubes and caps may be frozen at temperatures as low as -86˚C. The maximum centrifugal force is 12,000xg for the 10ml or 15ml tubes and 15,000xg for the 50 ml tubes*. Caps have a flat top surface for easy marking and labeling. Cat. No. C2605 features a ‘skirted’ bottom that allows the tube to stand upright without a separate test tube rack.*Refers to liquids with a specific gravity of 1.0, using appropriate rotors, cushions, and adapters.
Catalog No. NC2017618 Supplier MTC Bio Supplier No. C2410
Conical 10ml, 15ml, and 50ml centrifuge tubes are made of premium polypropylene for clarity and centrifugal strength. Tubes and caps may be frozen at temperatures as low as -86˚C. The maximum centrifugal force is 12,000xg for the 10ml or 15ml tubes and 15,000xg for the 50 ml tubes*. Caps have a flat top surface for easy marking and labeling. Cat. No. C2605 features a ‘skirted’ bottom that allows the tube to stand upright without a separate test tube rack.*Refers to liquids with a specific gravity of 1.0, using appropriate rotors, cushions, and adapters.